  • Search Results
  • Toll-like receptor 9 activation is a key mechanism for the maintenance of chronic lung inflammation.
Toll-like receptor 9 activation is a key mechanism for the maintenance of chronic lung inflammation.
American journal of respiratory and critical care medicine (2009-10-03)
Toshihiro Ito, Matthew Schaller, Tracy Raymond, Amrita D Joshi, Ana L Coelho, Fabiani G Frantz, William F Carson, Cory M Hogaboam, Nicholas W Lukacs, Theodore J Standiford, Sem H Phan, Stephen W Chensue, Steven L Kunkel
ABSTRACT
Accumulating evidence supports the hypothesis that the continuous host response to a persistent challenge can polarize the cytokine environment toward a Th2 cytokine phenotype, but the mechanisms responsible for this skewing are not clear. We investigated the role of Toll-like receptor 9 (TLR9) in a Th2-driven pulmonary granulomatous response initiated via the embolization of Schistosoma mansoni eggs to the lungs of mice. Mice were intravenously injected with S. mansoni eggs. Histological and flow cytometric analysis, cytokine measurement, adoptive transfer of bone marrow (BM)-derived dendritic cells (DCs), and in vitro T-cell treatments with antigen-presenting cells were examined. In comparison to wild-type mice, TLR9(-/-) mice showed increased pulmonary granuloma size, augmented collagen deposition, increased Th2 cytokine phenotype, and impaired accumulation of DCs. BM-derived DCs, but not macrophages, recovered from animals with developed Th2-type lung granulomas promoted the production of type 2 cytokines from CD4(+) T cells. BM-derived DCs from TLR9(-/-) mice induced impaired Th1 cytokine and enhanced Th2 cytokine production by T cells, compared with DCs from WT mice. Macrophages from TLR9(-/-) mice expressed a significantly higher alternatively activated (M2) phenotype characterized by increased "found in inflammatory zone-1" (FIZZ1) and arginase-1 expression. The adoptive transfer of BM-derived DCs from syngeneic WT mice into TLR9(-/-) mice restored the granuloma phenotype seen in WT mice. These studies suggest that TLR9 plays an important mechanistic role in the maintenance of the pulmonary granulomatous response.
Product Description
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture