  • Search Results
  • Conditional ablation of MAPK7 expression in chondrocytes impairs endochondral bone formation in limbs and adaptation of chondrocytes to hypoxia.
Conditional ablation of MAPK7 expression in chondrocytes impairs endochondral bone formation in limbs and adaptation of chondrocytes to hypoxia.
Cell & bioscience (2020-09-19)
Xiaoming Yang, Dongmei Zhong, Wenjie Gao, Zhiheng Liao, Yuyu Chen, Shun Zhang, Hang Zhou, Peiqiang Su, Caixia Xu
ABSTRACT
Long bones of limbs are formed through endochondral bone formation, which depends on the coordinated development of growth plates. Our previous studies have demonstrated that dysfunction of mitogen-activated protein kinase 7 (MAPK7) can cause skeletal dysplasia. However, little is known about the role of MAPK7 in the regulation of proliferation and differentiation of chondrocytes during growth plate development. Ablation of MAPK7 expression in chondrocytes led to growth restriction, short limbs and bone mass loss in postnatal mice. Histological studies revealed that MAPK7 deficiency increased the apoptosis and decreased the proliferation of chondrocytes in the center of the proliferative layer, where the most highly hypoxic chondrocytes are located. Accordingly, hypertrophic differentiation markers were downregulated in the central hypertrophic layer, beneath the site where abnormal apoptosis was observed. Simultaneously, we demonstrated that hypoxic adaptation and hypoxia-induced activation of hypoxia-inducible factor 1 subunit α (HIF1α) were impaired when MAPK7 could not be activated normally in primary chondrocytes. Concomitantly, vascular invasion into epiphyseal cartilage was inhibited when Mapk7 was deleted. We demonstrated that MAPK7 is necessary for maintaining proliferation, survival, and differentiation of chondrocytes during postnatal growth plate development, possibly through modulating HIF1α signaling for adaptation to hypoxia. These results indicate that MAPK7 signaling might be a target for treatment of chondrodysplasia.
Sigma-Aldrich
ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Alizarin Red S, certified by the Biological Stain Commission
Sigma-Aldrich
BCIP®/NBT-Purple Liquid Substrate System for Membranes, ready to use alkaline phosphatase substrate
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)