  • Search Results
  • HIV-1 gp41-specific monoclonal mucosal IgAs derived from highly exposed but IgG-seronegative individuals block HIV-1 epithelial transcytosis and neutralize CD4(+) cell infection: an IgA gene and functional analysis.
HIV-1 gp41-specific monoclonal mucosal IgAs derived from highly exposed but IgG-seronegative individuals block HIV-1 epithelial transcytosis and neutralize CD4(+) cell infection: an IgA gene and functional analysis.
Mucosal immunology (2009-07-10)
D Tudor, M Derrien, L Diomede, A-S Drillet, M Houimel, C Moog, J-M Reynes, L Lopalco, M Bomsel
ABSTRACT
AIDS is mainly a sexually transmitted disease, and accordingly, mucosal tissues are the primary sites of natural human immunodeficiency virus type-1 (HIV-1) transmission. Mucosal immunoglobulin A (IgA) antibody specific for HIV-1 envelope gp41 subunit is one correlate of protection in individuals who are highly sexually exposed to HIV-1 but remain persistently IgG seronegative (HEPS). Understanding these peculiar IgAs at the gene and functional level is possible only with monoclonal IgAs. We have constructed a mucosal Fab IgA library from HEPS and have characterized a series of HIV-1 IgAs specific for gp41 that, in vitro, are transcytosis-blocking and infection-neutralizing. Characterization of their IgA genes shows that Fab specific for the gp41 membrane-proximal region harbors a long heavy-chain CDR3 loop (CDRH3) similar to the two broadly neutralizing IgG monoclonal antibodies, 2F5 and 4E10. Furthermore, the selected Fab IgA shows extensive somatic mutations that cluster in the CDR regions, indicating that affinity maturation due to an antigen-driven process had occurred in HEPS individuals, presumably upon multiple exposures to HIV. This analysis of HEPS monoclonal IgA gives a unique opportunity to correlate an antibody function (resistance to a pathogen in vivo) with an antibody gene. Such neutralizing monoclonal IgAs could be used in microbicide formulation.
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution
Sigma-Aldrich
IgA from human colostrum, reagent grade, buffered aqueous solution