  • Search Results
  • Comparison of the Antiaging and Protective Properties of Plants from the Apiaceae Family.
Comparison of the Antiaging and Protective Properties of Plants from the Apiaceae Family.
Oxidative medicine and cellular longevity (2020-09-24)
Nizioł-Łukaszewska Zofia, Zagórska-Dziok Martyna, Ziemlewska Aleksandra, Bujak Tomasz
ABSTRACT
Plant materials play a very significant role as components of products being used both for medicinal and cosmetic purposes. Due to the high content of active substances, they can play an important role as extracts with antioxidant, regenerative, and antiaging properties. The skin aging process depends on various pathological and physiological processes, among which the degradation of extracellular matrix biomolecules such as collagen and elastin, which significantly affect the maintenance of good skin condition, is very important. The secondary metabolites and plant extracts may have collagenase and elastase inhibitory activity. This activity is mainly due to the high content of a wide range of various biologically active compounds, such as polyphenols, which include, among others, flavonoids, phenolic acids, tocopherols, and tannins. The work involved a comprehensive assessment of the plant from Apiaceae family such as Meum athamanticum L., Centella asiatica L., and Aegopodium podagraria L. extract as a multifunctional raw material. During study antioxidant properties, phenolic compounds and flavonoids content, effect on collagenase and elastase enzyme activity (antiaging effect), cytotoxic properties on skin cells (keratinocytes and fibroblasts), and cell migration capacity were analyzed. It has been shown that the highest antioxidant capacity can be observed for the extract of herb of Aegopodium podagraria L. When the concentration reached 5% all tested extracts had a positive effect on the cell proliferation of both keratinocytes and fibroblasts. It turned out that the most promising inhibitor of collagenase and elastase enzymes was the extract from Aegopodium podagraria, which inhibits the activity of both enzymes by over 70% in the concentration of 5% positively affecting the condition of skin cells.
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Resazurin sodium salt, certified by the Biological Stain Commission