  • Search Results
  • Gene expression profiling in dorsolateral prostates of prepubertal and adult Sprague-Dawley rats dosed with estradiol benzoate, estradiol, and testosterone.
Gene expression profiling in dorsolateral prostates of prepubertal and adult Sprague-Dawley rats dosed with estradiol benzoate, estradiol, and testosterone.
The Journal of toxicological sciences (2020-08-04)
Noriko Nakamura, Vikrant Vijay, Daniel T Sloper
ABSTRACT
The imbalance of testosterone to estradiol ratio has been related to the development of prostate diseases. Although rat models of prostate diseases induced by endocrine-disrupting chemicals (EDCs) and/or hormone exposure are commonly used to analyze gene expression profiles in the prostate, most studies utilize a single endpoint. In this study, microarray analysis was used for gene expression profiling in rat prostate tissue after exposure to EDCs and sex hormones over multiple time points (prepubertal through adulthood). We used dorsolateral prostate tissues from Sprague-Dawley rats (male offspring) and postnatally administered estradiol benzoate (EB) on postnatal days (PNDs) 1, 3, and 5, followed by treatment with additional hormones [estradiol (E) and testosterone (T)] on PNDs 90-200, as described by Ho et al. Microarray analysis was performed for gene expression profiling in the dorsolateral prostate, and the results were validated via qRT-PCR. The genes in cytokine-cytokine receptor interaction, cell adhesion molecules, and chemokines were upregulated in the EB+T+E group on PNDs 145 and 200. Moreover, early-stage downregulation of anti-inflammatory gene: bone morphogenetic protein 7 gene was observed. These findings suggest that exposure to EB, T, and E activates multiple pathways and simultaneously downregulates anti-inflammatory genes. Interestingly, these genes are reportedly expressed in prostate cancer tissues/cell lines. Further studies are required to elucidate the mechanism, including analyses using human prostate tissues.
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%