  • Search Results
  • The Perspective of Diagnostic and Prognostic Values of Lipoxygenases mRNA Expression in Colon Adenocarcinoma.
The Perspective of Diagnostic and Prognostic Values of Lipoxygenases mRNA Expression in Colon Adenocarcinoma.
OncoTargets and therapy (2020-10-17)
Guo-Tian Ruan, Yi-Zhen Gong, Li-Chen Zhu, Feng Gao, Xi-Wen Liao, Xiang-Kun Wang, Guang-Zhi Zhu, Cun Liao, Shuai Wang, Ling Yan, Hai-Lun Xie, Xin Zhou, Jun-Qi Liu, Meng-Nan Shao, Jia-Liang Gan
ABSTRACT
This study was mainly to explore and study the potential application of lipoxygenases (ALOX) family genes in the diagnostic and prognostic values of colon adenocarcinoma (COAD). Data sets related to the ALOX genes of COAD were obtained from The Cancer Genome Atlas and the University of California, Santa Cruz Xena browser. Then, the relevant biological information was downloaded from the public data platform. Finally, the bioinformatics technologies and clinical verification were employed to comprehensively analyze the potential values of ALOX genes. The Pearson correlation analysis indicated that there were correlations among ALOXE3, ALOX5, ALOX12, and ALOX12B. The diagnostic receiver operating characteristic (ROC) curves suggested that ALOXE3 and ALOX12 had significant diagnosis in COAD: ALOXE3; P<0.001, area under curve (AUC) 95%CI:=0.818 (0.773-0.862) and ALOX12; P<0.001, AUC 95%CI=0.774 (0.682-0.807). Besides, the verification study indicated that ALOX12 had a diagnostic value in COAD. Finally, our multivariate survival analysis and comprehensive prognosis of ALOX genes in COAD suggested that the ALOXE3 and ALOX12 were associated with COAD overall survival: ALOXE3; P=0.025, HR 95%CI=1.765 (1.074-2.901), ALOX12; P=0.046, HR 95%CI=1.680 (1.009-2.796), and the low expression of ALOXE3 and ALOX12 had a favorable prognosis of COAD (all P<0.05); on the contrary, the high regulation of them increased the risk of death. In our study, we observed that the mRNA expressions of ALOX genes were associated with the diagnosis and prognosis of COAD. The results of the diagnostic analysis suggested that ALOX12 might have a diagnosis value in COAD. Besides, our comprehensive prognosis analysis indicated that ALOXE3 combined ALOX12 might serve as potential prognosis biomarkers for COAD.
Sigma-Aldrich
12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ≥95% (HPLC)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.