  • Search Results
  • Pharmacological characteristics and binding modes of caracurine V analogues and related compounds at the neuronal alpha7 nicotinic acetylcholine receptor.
Pharmacological characteristics and binding modes of caracurine V analogues and related compounds at the neuronal alpha7 nicotinic acetylcholine receptor.
Journal of medicinal chemistry (2007-08-29)
Anders A Jensen, Darius P Zlotos, Tommy Liljefors
ABSTRACT
The pharmacological properties of bisquaternary caracurine V, iso-caracurine V, and pyrazino[1,2-a;4,5-a']diindole analogues and of the neuromuscular blocking agents alcuronium and toxiferine I have been characterized at numerous ligand-gated ion channels. Several of the analogues are potent antagonists of the homomeric alpha7 nicotinic acetylcholine receptor (nAChR), displaying nanomolar binding affinities and inhibiting acetylcholine-evoked signaling through the receptor in a competitive manner. In contrast, they do not display activities at heteromeric neuronal nAChRs and only exhibit weak antagonistic activities at the related 5-HT3A serotonin receptor. In a mutagenesis study, five selected analogues have been demonstrated to bind to the orthosteric site of the alpha7 nAChR. The binding site of the compounds overlaps with that of the standard alpha7 antagonist methyllycaconitine, the binding of them being centered in a cation-pi interaction between the quaternary nitrogen atom of the ligand and the Trp149 residue in the receptor, with additional key contributions from other aromatic receptor residues such as Tyr188, Tyr195, and Trp55.
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
SAFC
Glycine
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
(−)-Nicotine solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard