  • Search Results
  • Using migrating cells as probes to illuminate features in live embryonic tissues.
Using migrating cells as probes to illuminate features in live embryonic tissues.
Science advances (2020-12-06)
Sargon Gross-Thebing, Lukasz Truszkowski, Daniel Tenbrinck, Héctor Sánchez-Iranzo, Carolina Camelo, Kim J Westerich, Amrita Singh, Paul Maier, Jonas Prengel, Pia Lange, Jan Hüwel, Fjedor Gaede, Ramona Sasse, Bart E Vos, Timo Betz, Maja Matis, Robert Prevedel, Stefan Luschnig, Alba Diz-Muñoz, Martin Burger, Erez Raz
ABSTRACT
The biophysical and biochemical properties of live tissues are important in the context of development and disease. Methods for evaluating these properties typically involve destroying the tissue or require specialized technology and complicated analyses. Here, we present a novel, noninvasive methodology for determining the spatial distribution of tissue features within embryos, making use of nondirectionally migrating cells and software we termed "Landscape," which performs automatized high-throughput three-dimensional image registration. Using the live migrating cells as bioprobes, we identified structures within the zebrafish embryo that affect the distribution of the cells and studied one such structure constituting a physical barrier, which, in turn, influences amoeboid cell polarity. Overall, this work provides a unique approach for detecting tissue properties without interfering with animal's development. In addition, Landscape allows for integrating data from multiple samples, providing detailed and reliable quantitative evaluation of variable biological phenotypes in different organisms.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Sigma-Aldrich
Agarose, low gelling temperature, Type VII-A