Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Prednisolone 21-acetate, ≥97%
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Triamcinolone
Sigma-Aldrich
Glipizide, solid
Sigma-Aldrich
Hydrocortisone 21-acetate, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone 21-acetate, ≥98% (HPLC)
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%
Supelco
Salicylic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Betamethasone, ≥98%
Sigma-Aldrich
Betamethasone, meets USP testing specifications
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder