  • Search Results
  • Evidence of escape of SARS-CoV-2 variant B.1.351 from natural and vaccine-induced sera.
Evidence of escape of SARS-CoV-2 variant B.1.351 from natural and vaccine-induced sera.
Cell (2021-03-18)
Daming Zhou, Wanwisa Dejnirattisai, Piyada Supasa, Chang Liu, Alexander J Mentzer, Helen M Ginn, Yuguang Zhao, Helen M E Duyvesteyn, Aekkachai Tuekprakhon, Rungtiwa Nutalai, Beibei Wang, Guido C Paesen, Cesar Lopez-Camacho, Jose Slon-Campos, Bassam Hallis, Naomi Coombes, Kevin Bewley, Sue Charlton, Thomas S Walter, Donal Skelly, Sheila F Lumley, Christina Dold, Robert Levin, Tao Dong, Andrew J Pollard, Julian C Knight, Derrick Crook, Teresa Lambe, Elizabeth Clutterbuck, Sagida Bibi, Amy Flaxman, Mustapha Bittaye, Sandra Belij-Rammerstorfer, Sarah Gilbert, William James, Miles W Carroll, Paul Klenerman, Eleanor Barnes, Susanna J Dunachie, Elizabeth E Fry, Juthathip Mongkolsapaya, Jingshan Ren, David I Stuart, Gavin R Screaton
ABSTRACT
The race to produce vaccines against severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) began when the first sequence was published, and this forms the basis for vaccines currently deployed globally. Independent lineages of SARS-CoV-2 have recently been reported: UK, B.1.1.7; South Africa, B.1.351; and Brazil, P.1. These variants have multiple changes in the immunodominant spike protein that facilitates viral cell entry via the angiotensin-converting enzyme-2 (ACE2) receptor. Mutations in the receptor recognition site on the spike are of great concern for their potential for immune escape. Here, we describe a structure-function analysis of B.1.351 using a large cohort of convalescent and vaccinee serum samples. The receptor-binding domain mutations provide tighter ACE2 binding and widespread escape from monoclonal antibody neutralization largely driven by E484K, although K417N and N501Y act together against some important antibody classes. In a number of cases, it would appear that convalescent and some vaccine serum offers limited protection against this variant.
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.