  • Search Results
  • In Vitro and In Vivo Biocompatibility Studies of a Cast and Coated Titanium Alloy.
In Vitro and In Vivo Biocompatibility Studies of a Cast and Coated Titanium Alloy.
Molecules (Basel, Switzerland) (2020-07-31)
Ursula Sommer, Stephan Laurich, Lucie de Azevedo, Katharina Viehoff, Sabine Wenisch, Ulrich Thormann, Volker Alt, Christian Heiss, Reinhard Schnettler
ABSTRACT
The biocompatibility of a cast porous and with a calcium titanate reaction layer functionalized titanium alloy (Ti-6Al-7Nb) was tested by means of cell culture, and a small (rat) and large animal (sheep) model. The uncoated titanium material served as a control. In-vitro tests included the validation of osteoblast-like cells attached to the surface of the material with scanning electron microscopy and immunofluorescence of cytoskeletal actin as well as their osteogenic development, the ability to mineralize, and their vitality. Following the in-vitro tests a small animal (rat) and big animal (sheep) model were accomplished by inserting a cylindrical titanium implant into a drill hole defect in the femoral condyle. After 7, 14, and 30 days (rat) and 6 months (sheep) the condyles were studied regarding histological and histomorphometrical characteristics. Uncoated and coated material showed a good biocompatibility both in cell culture and animal models. While the defect area in the rat is well consolidated after 30 days, the sheep show only little bone inside the implant after 6 months, possibly due to stress shielding. None of the executed methods indicated a statistically significant difference between coated and uncoated material.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Eukitt® Quick-hardening mounting medium, for microscopy
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.)
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
Sodium chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Rat serum
Sigma-Aldrich
Alizarin-3-methyliminodiacetic acid
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Naphthol AS-TR phosphate disodium salt, ≥99% (HPLC), Bulk package
Sigma-Aldrich
6-Benzylaminopurine hydrochloride, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)