  • Search Results
  • Fibronectin Patches as Anchoring Points for Force Sensing and Transmission in Human Induced Pluripotent Stem Cell-Derived Pericytes.
Fibronectin Patches as Anchoring Points for Force Sensing and Transmission in Human Induced Pluripotent Stem Cell-Derived Pericytes.
Stem cell reports (2020-05-30)
Olga Iendaltseva, Valeria V Orlova, Christine L Mummery, Erik H J Danen, Thomas Schmidt
ABSTRACT
Pericytes (PCs) have been reported to contribute to the mechanoregulation of the capillary diameter and blood flow in health and disease. How this is realized remains poorly understood. We designed several models representing basement membrane (BM) in between PCs and endothelial cells (ECs). These models captured a unique protein organization with micron-sized FN patches surrounded by laminin (LM) and allowed to obtain quantitative information on PC morphology and contractility. Using human induced pluripotent stem cell-derived PCs, we could address mechanical aspects of mid-capillary PC behavior in vitro. Our results showed that PCs strongly prefer FN patches over LM for adhesion formation, have an optimal stiffness for spreading in the range of EC rigidity, and react in a non-canonical way with increased traction forces and reduced spreading on other stiffness then the optimal. Our approach opens possibilities to further study PC force regulation under well-controlled conditions.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid
Sigma-Aldrich
Anti-Integrin αV Antibody, clone LM142, ascites fluid, clone LM142, Chemicon®