  • Search Results
  • Galactosaminogalactan activates the inflammasome to provide host protection.
Galactosaminogalactan activates the inflammasome to provide host protection.
Nature (2020-12-04)
Benoit Briard, Thierry Fontaine, Parimal Samir, David E Place, Laetitia Muszkieta, R K Subbarao Malireddi, Rajendra Karki, Shelbi Christgen, Perrine Bomme, Peter Vogel, Rémi Beau, Emilia Mellado, Oumaima Ibrahim-Granet, Bernard Henrissat, Ravi C Kalathur, Cam Robinson, Jean-Paul Latgé, Thirumala-Devi Kanneganti
ABSTRACT
Inflammasomes are important sentinels of innate immune defence that are activated in response to diverse stimuli, including pathogen-associated molecular patterns (PAMPs)1. Activation of the inflammasome provides host defence against aspergillosis2,3, which is a major health concern for patients who are immunocompromised. However, the Aspergillus fumigatus PAMPs that are responsible for inflammasome activation are not known. Here we show that the polysaccharide galactosaminogalactan (GAG) of A. fumigatus is a PAMP that activates the NLRP3 inflammasome. The binding of GAG to ribosomal proteins inhibited cellular translation machinery, and thus activated the NLRP3 inflammasome. The galactosamine moiety bound to ribosomal proteins and blocked cellular translation, which triggered activation of the NLRP3 inflammasome. In mice, a GAG-deficient Aspergillus mutant (Δgt4c) did not elicit protective activation of the inflammasome, and this strain exhibited enhanced virulence. Moreover, administration of GAG protected mice from colitis induced by dextran sulfate sodium in an inflammasome-dependent manner. Thus, ribosomes connect the sensing of this fungal PAMP to the activation of an innate immune response.
Sigma-Aldrich
Saponin, for molecular biology, used as non-ionic surfactant
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Roche
DOTAP Liposomal Transfection Reagent, >99% (TLC), liquid, suitable for transfection
Sigma-Aldrich
Cortisone 21-acetate, ≥99%
Sigma-Aldrich
Curdlan from Alcaligenes faecalis
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.