  • Search Results
  • Detection of Embryonic Suspensor Cell Death by Whole-Mount TUNEL Assay in Tobacco.
Detection of Embryonic Suspensor Cell Death by Whole-Mount TUNEL Assay in Tobacco.
Plants (Basel, Switzerland) (2020-09-17)
Ce Shi, Pan Luo, Peng Zhao, Meng-Xiang Sun
ABSTRACT
Embryonic suspensor in angiosperms is a short-lived structure that connects the embryo to surrounding maternal tissues, which is necessary for early embryogenesis. Timely degeneration via programed cell death is the most distinct feature of the suspensor during embryogenesis. Therefore, the molecular mechanism regulating suspensor cell death is worth in-depth study for embryonic development. However, this process can hardly be detected using conventional methods since early embryos are deeply embedded in the seed coats and inaccessible through traditional tissue section. Hence, it is necessary to develop a reliable protocol for terminal deoxynucleotidyl transferase (TdT) dUTP Nick-End Labeling (TUNEL) analysis using limited living early embryos. Here, we provide a detailed protocol for the whole-mount detection of suspensor cell death using a TUNEL system in tobacco. This method is especially useful for the direct and rapid detection of the spatial-temporal characters of programed cell death during embryogenesis, as well as for the diminishment of the artifacts during material treatment by traditional methods.
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Mineral oil, BioReagent, for molecular biology, light oil
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests