  • Search Results
  • Response of rainbow trout (Oncorhynchus mykiss) in skin and fin tissue during infection with a variant of Gyrodactylus salaris (Monogenea: Gyrodactylidae).
Response of rainbow trout (Oncorhynchus mykiss) in skin and fin tissue during infection with a variant of Gyrodactylus salaris (Monogenea: Gyrodactylidae).
Folia parasitologica (2010-02-05)
Thomas R Jørgensen, Martin K Raida, Per W Kania, Kurt Buchmann
ABSTRACT
Response mechanisms of rainbow trout Oncorhynchus mykiss (Walbaum), experimentally infected with a Danish strain of Gyrodactylus salaris Malmberg, 1957 were investigated using molecular tools (qPCR) and immunohistochemistry. Expression of ten immune-relevant genes and reactivity with five different antibodies in the epidermis of skin and fin tissue were analysed in susceptible but responding rainbow trout. Rainbow trout were susceptible with regard to the parasite strain which initially colonised fins but relocated to the body region as infection progressed. The ten investigated genes encoding the cytokines IL-1beta, TNF-alpha, IFN-gamma, IL-10 and markers for adaptive immune activity, such as CD-4, CD-8, TCR-alpha, IgM, IgT and MHC II, were not found significantly regulated during the course of infection although IFN-gamma showed a slight up-regulation. Immunohistochemical analyses showed positive reactivity with antibodies against CD3, B-lymphocytes, neutrophilic granulocytes and collectin but not with mAb against IgM. No staining differences between infected and non-infected skin and fin tissue were detected.
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 350 purifications