  • Search Results
  • Modulation of dopamine D1 receptors via histamine H3 receptors is a novel therapeutic target for Huntington's disease.
Modulation of dopamine D1 receptors via histamine H3 receptors is a novel therapeutic target for Huntington's disease.
eLife (2020-06-10)
David Moreno-Delgado, Mar Puigdellívol, Estefanía Moreno, Mar Rodríguez-Ruiz, Joaquín Botta, Paola Gasperini, Anna Chiarlone, Lesley A Howell, Marco Scarselli, Vicent Casadó, Antoni Cortés, Sergi Ferré, Manuel Guzmán, Carmen Lluís, Jordi Alberch, Enric I Canela, Silvia Ginés, Peter J McCormick
ABSTRACT
Early Huntington's disease (HD) include over-activation of dopamine D1 receptors (D1R), producing an imbalance in dopaminergic neurotransmission and cell death. To reduce D1R over-activation, we present a strategy based on targeting complexes of D1R and histamine H3 receptors (H3R). Using an HD mouse striatal cell model and HD mouse organotypic brain slices we found that D1R-induced cell death signaling and neuronal degeneration, are mitigated by an H3R antagonist. We demonstrate that the D1R-H3R heteromer is expressed in HD mice at early but not late stages of HD, correlating with HD progression. In accordance, we found this target expressed in human control subjects and low-grade HD patients. Finally, treatment of HD mice with an H3R antagonist prevented cognitive and motor learning deficits and the loss of heteromer expression. Taken together, our results indicate that D1R - H3R heteromers play a pivotal role in dopamine signaling and represent novel targets for treating HD.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2, ascites fluid, clone 5TF1-1C2, Chemicon®
Sigma-Aldrich
Thioperamide maleate salt, solid
Sigma-Aldrich
Anti-Spinophilin/Neurabin-II Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-β-tubulin antibody, Mouse monoclonal, clone 2-28-33, purified from hybridoma cell culture