  • Search Results
  • LINE1 CpG-DNA Hypomethylation in Granulosa Cells and Blood Leukocytes Is Associated With PCOS and Related Traits.
LINE1 CpG-DNA Hypomethylation in Granulosa Cells and Blood Leukocytes Is Associated With PCOS and Related Traits.
The Journal of clinical endocrinology and metabolism (2017-03-23)
Pooja Sagvekar, Vijay Mangoli, Sadhana Desai, Anushree Patil, Srabani Mukherjee
ABSTRACT
Altered global DNA methylation is indicative of epigenomic instability concerning chronic diseases. Investigating its incidence and association with polycystic ovary syndrome (PCOS) is essential to understand the etiopathogenesis of this disorder. We assessed global DNA methylation differences in peripheral blood leukocytes (PBLs) and cumulus granulosa cells (CGCs) of controls and women with PCOS; and their association with PCOS and its traits. This study included a total of 102 controls and women with PCOS. Forty-one women undergoing controlled ovarian hyperstimulation (COH) and 61 women not undergoing COH were recruited from in vitro fertilization (IVF) and infertility clinics. DNA methylation was measured by ELISA for 5'-methyl-cytosine content and bisulfite sequencing of 5'-untranslated region (5'-UTR) of long interspersed nucleotide element-1 (LINE1/L1). Total 5'-methyl-cytosine and L1 methylation levels in PBLs and CGCs were similar between controls and women with PCOS. Methylation assessed at CpG sites of L1 5'-UTR revealed a single CpG-site (CpG-4) to be consistently hypomethylated in PBLs of both PCOS groups and CGCs of stimulated PCOS group. In unstimulated women, hypomethylation at CpG-4 was strongly associated with PCOS susceptibility, whereas in stimulated group it showed strong associations with PCOS and its hormonal traits. Furthermore, CGCs demonstrated consistent global and CpG-DNA hypomethylation relative to PBLs, irrespective of normal or disease states. Our study revealed strong association of single hypomethylated CpG-site with PCOS. Identification and characterization of more such methyl-CpG signatures in repetitive elements in larger study populations would provide valuable epigenetic insights into PCOS.
Sigma-Aldrich
Imprint® Methylated DNA Quantification Kit, To measure global DNA methylation shifts from as low as 10 ng DNA