  • Search Results
  • MHC/class-II-positive cells inhibit corticosterone of adrenal gland cells in experimental arthritis: a role for IL-1β, IL-18, and the inflammasome.
MHC/class-II-positive cells inhibit corticosterone of adrenal gland cells in experimental arthritis: a role for IL-1β, IL-18, and the inflammasome.
Scientific reports (2020-10-15)
Hubert Stangl, Anita Krammetsvogl, Martin Lesiak, Christine Wolff, Rainer H Straub
ABSTRACT
In experimental arthritis, glucocorticoid secretion is inadequate relative to inflammation. We hypothesized that IL-1 is a key factor for inadequate glucocorticoid secretion in arthritic rats. Collagen type II-induced arthritis (CIA) in DA rats was the model to study effects of IL-1 on adrenal function. In the CIA model, an increase of intraadrenal MHCII-positive cells was observed. MHCII-positive cells or bone marrow-derived dendritic cells inhibited glucocorticoid secretion of adrenal gland cells. IL-1, but also IL-18 and the inflammasome were critical in glucocorticoid inhibition. Arthritic compared to control adrenal gland cells produced higher amounts of CXC chemokines from MHCII+ adrenal cells, particularly CINC-2, which is strongly dependent on presence of IL-1. In CIA, macrophages and/or dendritic cells inhibit glucocorticoid secretion via IL-1 in adrenal glands. These findings show that activated macrophages and/or dendritic cells inhibit glucocorticoid secretion in experimental arthritis and that IL-1β is a decisive factor.
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, 0.25-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture