  • Search Results
  • Simultaneous quantification of protein-DNA interactions and transcriptomes in single cells with scDam&T-seq.
Simultaneous quantification of protein-DNA interactions and transcriptomes in single cells with scDam&T-seq.
Nature protocols (2020-05-01)
Corina M Markodimitraki, Franka J Rang, Koos Rooijers, Sandra S de Vries, Alex Chialastri, Kim L de Luca, Silke J A Lochs, Dylan Mooijman, Siddharth S Dey, Jop Kind
ABSTRACT
Protein-DNA interactions are essential for establishing cell type-specific chromatin architecture and gene expression. We recently developed scDam&T-seq, a multi-omics method that can simultaneously quantify protein-DNA interactions and the transcriptome in single cells. The method effectively combines two existing methods: DNA adenine methyltransferase identification (DamID) and CEL-Seq2. DamID works through the tethering of a protein of interest (POI) to the Escherichia coli DNA adenine methyltransferase (Dam). Upon expression of this fusion protein, DNA in proximity to the POI is methylated by Dam and can be selectively digested and amplified. CEL-Seq2, in contrast, makes use of poly-dT primers to reverse transcribe mRNA, followed by linear amplification through in vitro transcription. scDam&T-seq is the first technique capable of providing a combined readout of protein-DNA contact and transcription from single-cell samples. Once suitable cell lines have been established, the protocol can be completed in 5 d, with a throughput of hundreds to thousands of cells. The processing of raw sequencing data takes an additional 1-2 d. Our method can be used to understand the transcriptional changes a cell undergoes upon the DNA binding of a POI. It can be performed in any laboratory with access to FACS, robotic and high-throughput-sequencing facilities.
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Roche
Tris hydrochloride
Sigma-Aldrich
Mineral oil, light oil, BioReagent, suitable for mouse embryo cell culture
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%