CRISPR-Cas9-Mediated Genomic Deletions Protocol in Zebrafish.
STAR protocols (2020-12-31)
João Pedro Amorim, Renata Bordeira-Carriço, Ana Gali-Macedo, Chiara Perrod, José Bessa
ABSTRACT
Since its first application for site-directed mutagenesis, the CRISPR-Cas9 system has revolutionized genome engineering. Here, we present a validated workflow for the generation of targeted genomic deletions in zebrafish, including the design, cloning, and synthesis of single-guide RNAs and Cas9 mRNA, followed by microinjection in zebrafish embryos and subsequent genotype screening for the establishment of a mutant line. The versatility and efficiency of this pipeline makes the generation of zebrafish models a widely used approach in functional genetics. For complete details on the use and execution of this protocol, please refer to Amorim et al. (2020).
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Diethyl pyrocarbonate, ≥97% (NMR)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Yeast Extract, for use in microbial growth medium
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Phenol red solution, 0.5%, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%