  • Search Results
  • Glucose metabolism regulates expression of hair-inductive genes of dermal papilla spheres via histone acetylation.
Glucose metabolism regulates expression of hair-inductive genes of dermal papilla spheres via histone acetylation.
Scientific reports (2020-03-19)
Mina Choi, Yeong Min Choi, Soo-Young Choi, In-Sook An, Seunghee Bae, Sungkwan An, Jin Hyuk Jung
ABSTRACT
Cellular metabolism is one of the crucial factors to regulate epigenetic landscape in various cells including immune cells, embryonic stem cells and hair follicle stem cells. Dermal papilla cells (DP) interact with epithelial stem cells to orchestrate hair formation. Here we show that active DP exhibit robust aerobic glycolysis. We observed decrease of signature genes associated with hair induction by DP in presence of low glucose (2 mM) and glycolysis inhibitors. Moreover, hair shaft elongation was attenuated by glycolysis inhibitors. Interestingly, excessive glucose is able to increase the expression of hair inductive genes and elongation of hair shaft. We also observed glycolysis-mediated histone acetylation is increased and chemical inhibition of acetyltransferase reduces expression of the signature genes associated with hair induction in active DP. These results suggest that glucose metabolism is required for expression of signature genes associated with hair induction. This finding may be beneficial for establishing and maintaining of active DP to generate hair follicle in vitro.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Bromopyruvic acid, ≥98.0%
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
C646, ≥98% (HPLC)
Sigma-Aldrich
WZB-117, ≥98% (HPLC)