  • Search Results
  • Pentamidine inhibits prostate cancer progression via selectively inducing mitochondrial DNA depletion and dysfunction.
Pentamidine inhibits prostate cancer progression via selectively inducing mitochondrial DNA depletion and dysfunction.
Cell proliferation (2019-11-14)
Lin Liu, Fan Wang, Yu Tong, Lin-Feng Li, Yanfeng Liu, Wei-Qiang Gao
ABSTRACT
We investigated the anti-cancer activity of pentamidine, an anti-protozoal cationic aromatic diamidine drug, in prostate cancer cells and aimed to provide valuable insights for improving the efficacy of prostate cancer treatment. Prostate cancer cell lines and epithelial RWPE-1 cells were used in the study. Cell viability, wound-healing, transwell and apoptosis assays were examined to evaluate the influences of pentamidine in vitro. RNA-seq and qPCR were performed to analyse changes in gene transcription levels upon pentamidine treatment. Mitochondrial changes were assessed by measuring mitochondrial DNA content, morphology, membrane potential, cellular glucose uptake, ATP production and ROS generation. Nude mouse xenograft models were used to test anti-tumour effects of pentamidine in vivo. Pentamidine exerted profound inhibitory effects on proliferation, colony formation, migration and invasion of prostate cancer cells. In addition, the drug suppressed growth of xenograft tumours without exhibiting any obvious toxicity in nude mice. Mechanistically, pentamidine caused mitochondrial DNA content reduction and induced mitochondrial morphological changes, mitochondrial membrane potential dissipation, ATP level reduction, ROS production elevation and apoptosis in prostate cancer cells. Pentamidine can efficiently suppress prostate cancer progression and may serve as a novel mitochondria-targeted therapeutic agent for prostate cancer.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
2′,7′-Dichlorofluorescein diacetate, BioReagent, suitable for fluorescence, ≥95% (HPLC)