MilliporeSigma
  • A Non-Vector Approach to Increase Lipid Levels in the Microalga Planktochlorella nurekis.
A Non-Vector Approach to Increase Lipid Levels in the Microalga Planktochlorella nurekis.
Molecules (Basel, Switzerland) (2020-01-16)
Ewa Szpyrka, Daniel Broda, Bernadetta Oklejewicz, Magdalena Podbielska, Magdalena Slowik-Borowiec, Bartosz Jagusztyn, Grzegorz Chrzanowski, Malgorzata Kus-Liskiewicz, Magdalena Duda, Janusz Zuczek, Maciej Wnuk, Anna Lewinska
ABSTRACT
Microalgae are freshwater and marine unicellular photosynthetic organisms that utilize sunlight to produce biomass. Due to fast microalgal growth rate and their unique biochemical profiles and potential applications in food and renewable energy industries, the interest in microalgal research is rapidly increasing. Biochemical and genetic engineering have been considered to improve microalgal biomass production but these manipulations also limited microalgal growth. The aim of the study was the biochemical characterization of recently identified microalgal strain Planktochlorella nurekis with elevated cell size and DNA levels compared to wild type strain that was achieved by a safe non-vector approach, namely co-treatment with colchicine and cytochalasin B (CC). A slight increase in growth rate was observed in twelve clones of CC-treated cells. For biochemical profiling, several parameters were considered, namely the content of proteins, amino acids, lipids, fatty acids, β-glucans, chlorophylls, carotenoids, B vitamins and ash. CC-treated cells were characterized by elevated levels of lipids compared to unmodified cells. Moreover, the ratio of carotenoids to chlorophyll a and total antioxidant capacity were slightly increased in CC-treated cells. We suggest that Planktochlorella nurekis with modified DNA levels and improved lipid content can be considered to be used as a dietary supplement and biofuel feedstock.
Product Description
Supelco
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL
Pricing and availability is not currently available.