  • Search Results
  • Prediction of volume of distribution values in human using immobilized artificial membrane partitioning coefficients, the fraction of compound ionized and plasma protein binding data.
Prediction of volume of distribution values in human using immobilized artificial membrane partitioning coefficients, the fraction of compound ionized and plasma protein binding data.
European journal of medicinal chemistry (2009-07-10)
Xiaofan Sui, Jin Sun, Haiyan Li, Yongjun Wang, Jianfang Liu, Xiaohong Liu, Wenping Zhang, Lijiang Chen, Zhonggui He
ABSTRACT
We developed an improved Lombardeo's method (J. Med. Chem., 2004) for prediction of VD(ss) in human. With ElogD substituted by logk(IAM), together with f(i (7.4)) (the fraction of compound ionized at pH 7.4) and logf(u) (logarithmic fraction of compound unbound in plasma), the predictive equation of f(ut) (the fraction of the compound unbound in tissues) for the 121 compounds was built, predictive VD(ss) was further obtained from the Øie-Tozer equation. Our model could be applicable to structurally diverse compounds, including acids, bases, neutrals, and ampholytes. Interior and exterior validation results indicated the model had a robust predictive ability. Compared to the Lombardeo's and Hollósy's (J. Med. Chem., 2006) methods, our model can be generally applicable with better predictive accuracy.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Amoxicillin, potency: ≥900 μg per mg
Sigma-Aldrich
Trimethoprim, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Acetaminophen, analytical standard