  • Search Results
  • Diminution in sperm quantity and quality in mouse models of Duchenne Muscular Dystrophy induced by a myostatin-based muscle growth-promoting intervention.
Diminution in sperm quantity and quality in mouse models of Duchenne Muscular Dystrophy induced by a myostatin-based muscle growth-promoting intervention.
European journal of translational myology (2020-08-13)
Danielle Vaughan, Oliver Kretz, Ali Alqallaf, Robert Mitchell, Jennie L von der Heide, Sakthivel Vaiyapuri, Antonios Matsakas, Arja Pasternack, Henry Collins-Hooper, Olli Ritvos, Randy Ballesteros, Tobias B Huber, Helge Amthor, Abir Mukherjee, Ketan Patel
ABSTRACT
Duchenne Muscular Dystrophy is a devastating disease caused by the absence of a functional rod-shaped cytoplasmic protein called dystrophin. Several avenues are being developed aimed to restore dystrophin expression in boys affected by this X-linked disease. However, its complete cure is likely to need combinational approaches which may include regimes aimed at restoring muscle mass. Augmenting muscle growth through the manipulation of the Myostatin/Activin signalling axis has received much attention. However, we have recently shown that while manipulation of this axis in wild type mice using the sActRIIB ligand trap indeed results in muscle growth, it also had a detrimental impact on the testis. Here we examined the impact of administering a powerful Myostatin/Activin antagonist in two mouse models of Duchenne Muscular Dystrophy. We report that whilst the impact on muscle growth was not always positive, both models showed attenuated testis development. Sperm number, motility and ultrastructure were significantly affected by the sActRIIB treatment. Our report suggests that interventions based on Myostatin/Activin should investigate off-target effects on tissues as well as muscle.
Sigma-Aldrich
Mineral oil, light oil, BioReagent, suitable for mouse embryo cell culture
Sigma-Aldrich
Hematoxylin Solution, Harris Modified