  • Search Results
  • Surface texture limits transfer of S. aureus, T4 bacteriophage, influenza B virus and human coronavirus.
Surface texture limits transfer of S. aureus, T4 bacteriophage, influenza B virus and human coronavirus.
PloS one (2020-12-29)
Qi Liu, Lindsey Brookbank, Angela Ho, Jenna Coffey, Anthony B Brennan, Christopher J Jones
ABSTRACT
Spread of pathogens on contaminated surfaces plays a key role in disease transmission. Surface technologies that control pathogen transfer can help control fomite transmission and are of great interest to public health. Here, we report a novel bead transfer method for evaluating fomite transmission in common laboratory settings. We show that this method meets several important criteria for quantitative test methods, including reasonableness, relevancy, resemblance, responsiveness, and repeatability, and therefore may be adaptable for standardization. In addition, this method can be applied to a wide variety of pathogens including bacteria, phage, and human viruses. Using the bead transfer method, we demonstrate that an engineered micropattern limits transfer of Staphylococcus aureus by 97.8% and T4 bacteriophage by 93.0% on silicone surfaces. Furthermore, the micropattern significantly reduces transfer of influenza B virus and human coronavirus on silicone and polypropylene surfaces. Our results highlight the potential of using surface texture as a valuable new strategy in combating infectious diseases.
Sigma-Aldrich
LB Broth (Lennox), Highly-referenced microbial growth powder medium, low salt, suitable for salt-sensitive E.coli culture.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Crystal violet solution, 1%, aqueous solution