  • Search Results
  • A Novel Microfluidic Method Utilizing a Hydrofoil Structure to Improve Circulating Tumor Cell Enrichment: Design and Analytical Validation.
A Novel Microfluidic Method Utilizing a Hydrofoil Structure to Improve Circulating Tumor Cell Enrichment: Design and Analytical Validation.
Micromachines (2020-11-05)
Gürhan Özkayar, Ege Mutlu, Şebnem Şahin, Yağmur Demircan Demircan Yalçın, Taylan Töral, Haluk Külah, Ender Yıldırım, Özge Zorlu, Ebru Özgür
ABSTRACT
Being one of the major pillars of liquid biopsy, isolation and characterization of circulating tumor cells (CTCs) during cancer management provides critical information on the evolution of cancer and has great potential to increase the success of therapies. In this article, we define a novel strategy to effectively enrich CTCs from whole blood based on size, utilizing a spiral microfluidic channel embedded with a hydrofoil structure at the downstream of the spiral channel. The hydrofoil increases the distance between the streams of CTCs and peripheral blood cells, which are already distributed about two focal axes by the spiral channel, thereby improving the resolution of the separation. Analytical validation of the system has been carried out using Michigan Cancer Foundation-7 (MCF7) breast cancer cell lines spiked into blood samples from healthy donors, and the performance of the system in terms of white blood cell (WBC) depletion, CTC recovery rate and cell viability has been shown in single or two-step process: by passing the sample once or twice through the microfluidic chip. Single step process yielded high recovery (77.1%), viable (84.7%) CTCs. When the collected cell suspension is re-processed by the same chip, recovery decreases to 65.5%, while the WBC depletion increases to 88.3%, improving the purity. Cell viability of >80% was preserved after two-step process. The novel microfluidic chip is a good candidate for CTC isolation applications requiring high recovery rate and viability, including functional downstream analyses for variety of cancer types.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA • 4Na per liter of Hanks′ Balanced Salt Solution with phenol red