  • Search Results
  • Smartphone-interrogated test supports for the enzymatic determination of putrescine and cadaverine in food.
Smartphone-interrogated test supports for the enzymatic determination of putrescine and cadaverine in food.
Analytical and bioanalytical chemistry (2020-05-14)
Isabel Sanz-Vicente, Ángel López-Molinero, Susana de Marcos, Jesús Navarro, Pablo Cebrián, Chantal Arruego, Victor Visiedo, Javier Galbán
ABSTRACT
Diamino-oxidase (DAO), horseradish peroxidase (HRP), and tetramethylbenzidine (TMB) have been immobilized into cellulose to obtain circular cellulose test supports (CCTSs) for the determination of cadaverine (Cad) and putrescine (Put). During the enzymatic reaction, TMB is oxidized and a blue spot is obtained. This color (RGB coordinates) is measured with a smartphone and a commercial application. The highest sensitivity is provided by the component R and a linear response is observed for low biogenic amine (BA) concentrations, but a second-order polynomial response better fits the experimental results for a wider concentration range. This has been successfully explained with a model developed to explain the RGB values obtained in this type of analytical system. Optimization studies enable CCTSs to be obtained for Put and Cad determination, which could be used (kept at 4 °C) for at least 45 days if a stabilizer (StabilCoat™ or StabilGuard™) is added during its synthesis. In these conditions, the R coordinate follows the model up to at least 4 × 10-4 M Put and/or Cad (both analytes give the same response). The method permits the Put and Cad determination from 5 × 10-5 M up to 4 × 10-4 M (RSD = 3%, n = 3). The CCTSs have been applied to Put + Cad determination in a tuna sample without any interference by other biogenic amines. The concentration found statistically agrees with that obtained using a HPLC-MS-validated method. Graphical abstract.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Peroxidase from horseradish, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
Putrescine dihydrochloride, ≥98% (TLC)
Sigma-Aldrich
2-Phenylethylamine hydrochloride, ≥98%
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Tryptamine hydrochloride, 99%