  • Search Results
  • Cytotoxic effects of cardiac glycosides in colon cancer cells, alone and in combination with standard chemotherapeutic drugs.
Cytotoxic effects of cardiac glycosides in colon cancer cells, alone and in combination with standard chemotherapeutic drugs.
Journal of natural products (2009-11-10)
Jenny Felth, Linda Rickardson, Josefin Rosén, Malin Wickström, Mårten Fryknäs, Magnus Lindskog, Lars Bohlin, Joachim Gullbo
ABSTRACT
Cardiac glycosides have been reported to exhibit cytotoxic activity against several different cancer types, but studies against colorectal cancer are lacking. In a screening procedure aimed at identifying natural products with activity against colon cancer, several cardiac glycosides were shown to be of interest, and five of these were further evaluated in different colorectal cancer cell lines and primary cells from patients. Convallatoxin (1), oleandrin (4), and proscillaridin A (5) were identified as the most potent compounds (submicromolar IC50 values), and digitoxin (2) and digoxin (3), which are used in cardiac disease, exhibited somewhat lower activity (IC50 values 0.27-4.1 microM). Selected cardiac glycosides were tested in combination with four clinically relevant cytotoxic drugs (5-fluorouracil, oxaliplatin, cisplatin, irinotecan). The combination of 2 and oxaliplatin exhibited synergism including the otherwise highly drug-resistant HT29 cell line. A ChemGPS-NP application comparing modes of action of anticancer drugs identified cardiac glycosides as a separate cluster. These findings demonstrate that such substances may exhibit significant activity against colorectal cancer cell lines, by mechanisms disparate from currently used anticancer drugs, but at concentrations generally considered not achievable in patient plasma.
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder
Supelco
Oleandrin, analytical standard
Supelco
Digoxin, analytical standard
Sigma-Aldrich
Convallatoxin, ≥65%
Supelco
Digoxin, analytical standard