  • Search Results
  • Oxidative stability, proteolysis, and in vitro digestibility of fresh and long-term frozen stored in-bag dry-aged lean beef.
Oxidative stability, proteolysis, and in vitro digestibility of fresh and long-term frozen stored in-bag dry-aged lean beef.
Food chemistry (2020-12-04)
Renyu Zhang, Michelle J Y Yoo, Mustafa M Farouk
ABSTRACT
Effect of air velocities, stepwise in-bag ageing and ageing time on the oxidative stability, proteolysis and digestibility of fresh and long-term frozen-stored dry-aged lean beef were studied. Increased air velocities and stepwise ageing regime had no effect (P > 0.05) on dry-aged beef lipid and protein oxidative stabilities and proteolysis pattern compared to control straight-dry-ageing. TBARS, protein carbonyl and free amino acids (FAAs) increased (P > 0.05) with ageing time. In-bag dry-ageing of beef improved its lipids and protein oxidative stability during long-term frozen storage compared to unaged beef. Improvement in beef protein digestibility was observed through increased release of FAAs and appearance of small protein fragments from SDS-PAGE in dry-aged samples compared to the unaged. The high lipid and protein oxidative stability of long-term frozen lean beef produced using stepwise in-bag ageing process suggest potential for the process to be used for producing dry-aged meat for export.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Supelco
Amino acid standards, physiological, analytical standard, acidics, neutrals, and basics