  • Search Results
  • Reengineering an Antiarrhythmic Drug Using Patient hiPSC Cardiomyocytes to Improve Therapeutic Potential and Reduce Toxicity.
Reengineering an Antiarrhythmic Drug Using Patient hiPSC Cardiomyocytes to Improve Therapeutic Potential and Reduce Toxicity.
Cell stem cell (2020-09-16)
Wesley L McKeithan, Dries A M Feyen, Arne A N Bruyneel, Karl J Okolotowicz, Daniel A Ryan, Kevin J Sampson, Franck Potet, Alex Savchenko, Jorge Gómez-Galeno, Michelle Vu, Ricardo Serrano, Alfred L George, Robert S Kass, John R Cashman, Mark Mercola
ABSTRACT
Modeling cardiac disorders with human induced pluripotent stem cell (hiPSC)-derived cardiomyocytes is a new paradigm for preclinical testing of candidate therapeutics. However, disease-relevant physiological assays can be complex, and the use of hiPSC-cardiomyocyte models of congenital disease phenotypes for guiding large-scale screening and medicinal chemistry have not been shown. We report chemical refinement of the antiarrhythmic drug mexiletine via high-throughput screening of hiPSC-CMs derived from patients with the cardiac rhythm disorder long QT syndrome 3 (LQT3) carrying SCN5A sodium channel variants. Using iterative cycles of medicinal chemistry synthesis and testing, we identified drug analogs with increased potency and selectivity for inhibiting late sodium current across a panel of 7 LQT3 sodium channel variants and suppressing arrhythmic activity across multiple genetic and pharmacological hiPSC-CM models of LQT3 with diverse backgrounds. These mexiletine analogs can be exploited as mechanistic probes and for clinical development.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Sodium L-lactate, ≥99.0% (NT)
Sigma-Aldrich
L-Carnitine hydrochloride, synthetic, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Creatine monohydrate, ≥98%
Sigma-Aldrich
Ranolazine dihydrochloride, ≥98% (HPLC), powder