  • Search Results
  • Iron loading exerts synergistic action via a different mechanistic pathway from that of acetaminophen-induced hepatic injury in mice.
Iron loading exerts synergistic action via a different mechanistic pathway from that of acetaminophen-induced hepatic injury in mice.
Free radical research (2020-09-09)
Gyul Moon, Sho Kobayashi, Ye Aung Naing, Ken-Ichi Yamada, Mitsunori Yamakawa, Junichi Fujii
ABSTRACT
Acetaminophen (APAP) overdose is a major cause of drug-induced acute liver failure. In such cases, free iron is released from lysosomes and is transported to mitochondria where it plays a pivotal role in APAP-induced liver injury. We previously reported that ascorbic acid (Asc) markedly mitigates APAP-induced hepatic damage in aldehyde reductase (Akr1a)-knockout (KO) mice that produce about 10% Asc as wild-type (WT) mice. However, the issue of the protective mechanism of Asc in association with the status of iron remains ambiguous. To gain additional insights into this issue, we examined effects of APAP (500 mg/kg) on female KO mice under conditions of iron loading. While the KO mice without AsA supplementation were more sensitive to APAP toxicity than the WT mice, FeSO4 loading (25 mg/kg) to WT mice aggravated the hepatic injury, which was a similar extent to that of the KO mice. Supplementation of Asc (1.5 mg/ml in the drinking water) ameliorated KO mice irrespective of iron status but did not change the iron-mediated increase in the lethality in the WT mice. Hepatic cysteine and glutathione levels declined to similar extents in all mouse groups at 3 h irrespective of the iron status and largely recovered at 18 h after the APAP treatment when liver damage was evident. Asc prominently mitigated APAP toxicity in KO mice irrespective of the iron status but had no effect on the synergistic action of iron and APAP in the WT mice, suggesting that the mechanism for the deteriorating action of loaded iron is different from that of APAP toxicity.
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
N-Methylmaleimide, 97%