  • Search Results
  • Human neurotropic polyomavirus, JC virus, agnoprotein targets mitochondrion and modulates its functions.
Human neurotropic polyomavirus, JC virus, agnoprotein targets mitochondrion and modulates its functions.
Virology (2020-12-06)
Reshu Saxena, Sami Saribas, Pooja Jadiya, Dhanendra Tomar, Rafal Kaminski, John W Elrod, Mahmut Safak
ABSTRACT
JC virus encodes an important regulatory protein, known as Agnoprotein (Agno). We have recently reported Agno's first protein-interactome with its cellular partners revealing that it targets various cellular networks and organelles, including mitochondria. Here, we report further characterization of the functional consequences of its mitochondrial targeting and demonstrated its co-localization with the mitochondrial networks and with the mitochondrial outer membrane. The mitochondrial targeting sequence (MTS) of Agno and its dimerization domain together play major roles in this targeting. Data also showed alterations in various mitochondrial functions in Agno-positive cells; including a significant reduction in mitochondrial membrane potential, respiration rates and ATP production. In contrast, a substantial increase in ROS production and Ca2+ uptake by the mitochondria were also observed. Finally, findings also revealed a significant decrease in viral replication when Agno MTS was deleted, highlighting a role for MTS in the function of Agno during the viral life cycle.
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology
Supelco
(±)-cis-4,4′-Dimethylaminorex solution, 1.0 mg/mL (1% 1M HCl in Methanol), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin solution from bovine pancreas, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate solution, 100 mM, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Roche
Tris hydrochloride, >99% (titration), pH 7.0-9.0, suitable for FISH
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Rhodamine conjugate, Chemicon®, from goat
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Rhodamine conjugate, Chemicon®, from goat