  • Search Results
  • Development of imidazole alkanoic acids as mGAT3 selective GABA uptake inhibitors.
Development of imidazole alkanoic acids as mGAT3 selective GABA uptake inhibitors.
European journal of medicinal chemistry (2011-03-01)
Silke Hack, Babette Wörlein, Georg Höfner, Jörg Pabel, Klaus T Wanner
ABSTRACT
A new series of potential GABA uptake inhibitors starting from of 1H-imidazol-4-ylacetic acid with the carboxylic acid side chain originating from different positions and varying in length have been synthesized and tested for the inhibitory potency at the four GABA uptake transporters mGAT1-4 stably expressed in HEK cells. Further two bicyclic compounds with a rigidified carboxylic acid side chain were included in this study. The results of the biological tests indicated that most ω-imidazole alkanoic and alkenoic acid derivatives exhibit the highest potencies as GABA uptake inhibitors at mGAT3.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
β-Alanine, 99%
SAFC
Glycine
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
5-Aminovaleric acid, 97%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
β-Alanine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
4-Imidazoleacrylic acid, 99%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
7-Aminoheptanoic acid, 98%
Sigma-Aldrich
β-Alanine, ≥98%, FG