Zika virus (ZIKV) was discovered over 70 years ago in East Africa, but little is known about its circulation and pathogenesis there. We screened 327 plasma samples collected 2-12 months after febrile illness in Western and coastal Kenya (1993-2016) for binding and neutralizing antibodies to distinguish ZIKV and dengue virus (DENV) responses, which we found were common in coastal Kenya. Two cases had durable ZIKV-specific antibodies and 2 cases had ZIKV antibodies at similar levels as DENV antibodies. This suggests low-level ZIKV circulation in Kenya over 2 decades and sets a baseline for future surveillance efforts in East Africa.