  • Search Results
  • Detection of pseudorabies virus antibody in swine oral fluid using a serum whole-virus indirect ELISA.
Detection of pseudorabies virus antibody in swine oral fluid using a serum whole-virus indirect ELISA.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc (2020-05-27)
Ting-Yu Cheng, Alexandra Buckley, Albert Van Geelen, Kelly Lager, Alexandra Henao-Díaz, Korakrit Poonsuk, Pablo Piñeyro, David Baum, Ju Ji, Chong Wang, Rodger Main, Jeffrey Zimmerman, Luis Giménez-Lirola
ABSTRACT
We evaluated the detection of pseudorabies virus (PRV) antibodies in swine oral fluid. Oral fluid and serum samples were obtained from 40 pigs allocated to 4 treatment groups (10 pigs/group): negative control (NC); wild-type PRV inoculation (PRV 3CR Ossabaw; hereafter PRV); PRV vaccination (Ingelvac Aujeszky MLV; Boehringer Ingelheim; hereafter MLV); and PRV vaccination followed by PRV inoculation at 21 d post-vaccination (MLV-PRV). Using a serum PRV whole-virus indirect IgG ELISA (Idexx Laboratories) adapted to the oral fluid matrix, PRV antibody was detected in oral fluid samples from treatment groups PRV, MLV, and MLV-PRV in a pattern similar to serum. Vaccination alone produced a low oral fluid antibody response (groups MLV and MLV-PRV), but a strong anamnestic response was observed following challenge with wild-type virus (group PRV). Analyses of the oral fluid PRV indirect IgG ELISA results showed good binary diagnostic performance (area under ROC curve = 93%) and excellent assay repeatability (intra-class correlation coefficient = 99.3%). The demonstrable presence of PRV antibodies in swine oral fluids suggests the possible use of oral fluids in pseudorabies surveillance.
Sigma-Aldrich
Human IgG ELISA Kit, for serum, plasma