  • Search Results
  • Microfluidic device with brain extracellular matrix promotes structural and functional maturation of human brain organoids.
Microfluidic device with brain extracellular matrix promotes structural and functional maturation of human brain organoids.
Nature communications (2021-08-07)
Ann-Na Cho, Yoonhee Jin, Yeonjoo An, Jin Kim, Yi Sun Choi, Jung Seung Lee, Junghoon Kim, Won-Young Choi, Dong-Jun Koo, Weonjin Yu, Gyeong-Eon Chang, Dong-Yoon Kim, Sung-Hyun Jo, Jihun Kim, Sung-Yon Kim, Yun-Gon Kim, Ju Young Kim, Nakwon Choi, Eunji Cheong, Young-Joon Kim, Hyunsoo Shawn Je, Hoon-Chul Kang, Seung-Woo Cho
ABSTRACT
Brain organoids derived from human pluripotent stem cells provide a highly valuable in vitro model to recapitulate human brain development and neurological diseases. However, the current systems for brain organoid culture require further improvement for the reliable production of high-quality organoids. Here, we demonstrate two engineering elements to improve human brain organoid culture, (1) a human brain extracellular matrix to provide brain-specific cues and (2) a microfluidic device with periodic flow to improve the survival and reduce the variability of organoids. A three-dimensional culture modified with brain extracellular matrix significantly enhanced neurogenesis in developing brain organoids from human induced pluripotent stem cells. Cortical layer development, volumetric augmentation, and electrophysiological function of human brain organoids were further improved in a reproducible manner by dynamic culture in microfluidic chamber devices. Our engineering concept of reconstituting brain-mimetic microenvironments facilitates the development of a reliable culture platform for brain organoids, enabling effective modeling and drug development for human brain diseases.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
1,9-Dimethyl-Methylene Blue zinc chloride double salt, Dye content 80 %
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Triton X-100, laboratory grade