  • Search Results
  • Immunogenicity of standard and extended dosing intervals of BNT162b2 mRNA vaccine.
Immunogenicity of standard and extended dosing intervals of BNT162b2 mRNA vaccine.
Cell (2021-11-05)
Rebecca P Payne, Stephanie Longet, James A Austin, Donal T Skelly, Wanwisa Dejnirattisai, Sandra Adele, Naomi Meardon, Sian Faustini, Saly Al-Taei, Shona C Moore, Tom Tipton, Luisa M Hering, Adrienn Angyal, Rebecca Brown, Alexander R Nicols, Natalie Gillson, Susan L Dobson, Ali Amini, Piyada Supasa, Andrew Cross, Alice Bridges-Webb, Laura Silva Reyes, Aline Linder, Gurjinder Sandhar, Jonathan A Kilby, Jessica K Tyerman, Thomas Altmann, Hailey Hornsby, Rachel Whitham, Eloise Phillips, Tom Malone, Alexander Hargreaves, Adrian Shields, Ayoub Saei, Sarah Foulkes, Lizzie Stafford, Sile Johnson, Daniel G Wootton, Christopher P Conlon, Katie Jeffery, Philippa C Matthews, John Frater, Alexandra S Deeks, Andrew J Pollard, Anthony Brown, Sarah L Rowland-Jones, Juthathip Mongkolsapaya, Eleanor Barnes, Susan Hopkins, Victoria Hall, Christina Dold, Christopher J A Duncan, Alex Richter, Miles Carroll, Gavin Screaton, Thushan I de Silva, Lance Turtle, Paul Klenerman, Susanna Dunachie
ABSTRACT
Extension of the interval between vaccine doses for the BNT162b2 mRNA vaccine was introduced in the United Kingdom to accelerate population coverage with a single dose. At this time, trial data were lacking, and we addressed this in a study of United Kingdom healthcare workers. The first vaccine dose induced protection from infection from the circulating alpha (B.1.1.7) variant over several weeks. In a substudy of 589 individuals, we show that this single dose induces severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) neutralizing antibody (NAb) responses and a sustained B and T cell response to the spike protein. NAb levels were higher after the extended dosing interval (6-14 weeks) compared with the conventional 3- to 4-week regimen, accompanied by enrichment of CD4+ T cells expressing interleukin-2 (IL-2). Prior SARS-CoV-2 infection amplified and accelerated the response. These data on dynamic cellular and humoral responses indicate that extension of the dosing interval is an effective immunogenic protocol.
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
L-Glutamine–Penicillin–Streptomycin solution, with 200 mM L-glutamine, 10,000 U penicillin and 10 mg steptomycin/mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Roche
Phytohemagglutinin-L (PHA-L), from Phaseolus vulgaris