MilliporeSigma
Deciphering cellular signals in adult mouse sinoatrial node cells.
iScience (2022-01-18)
Gopireddy R Reddy, Lu Ren, Phung N Thai, Jessica L Caldwell, Manuela Zaccolo, Julie Bossuyt, Crystal M Ripplinger, Yang K Xiang, Madeline Nieves-Cintrón, Nipavan Chiamvimonvat, Manuel F Navedo
ABSTRACT
Sinoatrial node (SAN) cells are the pacemakers of the heart. This study describes a method for culturing and infection of adult mouse SAN cells with FRET-based biosensors that can be exploited to examine signaling events. SAN cells cultured in media with blebbistatin or (S)-nitro-blebbistatin retain their morphology, protein distribution, action potential (AP) waveform, and cAMP dynamics for at least 40 h. SAN cells expressing targeted cAMP sensors show distinct β-adrenergic-mediated cAMP pools. Cyclic GMP, protein kinase A, Ca2+/CaM kinase II, and protein kinase D in SAN cells also show unique dynamics to different stimuli. Heart failure SAN cells show a decrease in cAMP and cGMP levels. In summary, a reliable method for maintaining adult mouse SAN cells in culture is presented, which facilitates studies of signaling networks and regulatory mechanisms during physiological and pathological conditions.
Sigma-Aldrich
(−)-Isoproterenol (+)-bitartrate salt, powder
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%