  • Search Results
  • Epithelial to mesenchymal transition is activated in metastatic pheochromocytomas and paragangliomas caused by SDHB gene mutations.
Epithelial to mesenchymal transition is activated in metastatic pheochromocytomas and paragangliomas caused by SDHB gene mutations.
The Journal of clinical endocrinology and metabolism (2012-04-12)
Céline Loriot, Nelly Burnichon, Noémie Gadessaud, Laure Vescovo, Laurence Amar, Rossella Libé, Jérôme Bertherat, Pierre-François Plouin, Xavier Jeunemaitre, Anne-Paule Gimenez-Roqueplo, Judith Favier
ABSTRACT
Pheochromocytoma and paraganglioma are rare neural-crest-derived tumors. They are metastatic in 15% of cases, and the identification of a germline mutation in the SDHB gene is a predictive risk factor for malignancy and poor prognosis. To date, the link between SDHB mutations and malignancy is still missing. Epithelial to mesenchymal transition (EMT) is a developmental event, reactivated in cancer cells to promote cell mobility and invasiveness. The aim of this study was to address the participation of EMT in the metastatic evolution of pheochromocytoma/paraganglioma. Transcriptomic profiling of EMT was performed on 188 tumor samples, using a set of 94 genes implicated in this pathway. Activation of EMT was further confirmed at protein level by immunohistochemistry in a second set of 93 tumors. Hierarchical unsupervised classification showed that most SDHB-metastatic samples clustered together, indicating that EMT is differently regulated in these tumors. Major actors of EMT, metalloproteases and components of cellular junctions, were either up-regulated (LOXL2, TWIST, TCF3, MMP2, and MMP1) or down-regulated (KRT19 and CDH2) in SDHB-metastatic tumors compared with nonmetastatic ones. Interestingly, within metastatic tumors, most of these genes (LOXL2, TWIST, TCF3, MMP2, and KRT19) also allowed us to discriminate SDHB-mutated from non-SDHB-related tumors. In the second set of tumors, we studied Snail1/2 expression by immunohistochemistry and observed its specific nuclear translocation in all SDHB-metastatic tumors. We have identified the first pathway that distinguishes SDHB-metastatic from all other types of pheochromocytomas/paragangliomas and suggest that activation of the EMT process might play a critical role in the particularly invasive phenotype of this group of tumors.
Sigma-Aldrich
Anti-SDHB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution