  • Search Results
  • Pituitary adenylate cyclase activating polypeptide is an extraordinarily potent intra-pancreatic regulator of insulin secretion from islet beta-cells.
Pituitary adenylate cyclase activating polypeptide is an extraordinarily potent intra-pancreatic regulator of insulin secretion from islet beta-cells.
The Journal of biological chemistry (1994-01-14)
T Yada, M Sakurada, K Ihida, M Nakata, F Murata, A Arimura, M Kikuchi
ABSTRACT
Insulin secretion from pancreatic islets is controlled by peptides as well as by nutrients. We report here a novel, extraordinarily potent peptidergic regulation of insulin secretion. A 27-residue form of pituitary adenylate cyclase activating polypeptide (PACAP27) as low as 10(-14) to 10(-13) M stimulated insulin release from rat islets in a glucose-dependent manner. PACAP27 also increased cytosolic free Ca2+ concentration ([Ca2+]i) in islet beta-cells. Nitrendipine, a blocker of the L-type Ca2+ channel, abolished both [Ca2+]i and insulin responses. Vasoactive intestinal peptide, a peptide exhibiting 68% amino acid homology with PACAP, also increased [Ca2+]i in beta-cells but only at concentrations in the nanomolar range, indicating that PACAP27 is 4 logs more potent. A 38-residue form of the peptide (PACAP38) stimulated insulin release and increased beta-cell [Ca2+]i in a manner similar to that of PACAP27. PACAP-like immunoreactivity was demonstrated in pancreatic nerve fibers, islets, and capillaries. The results indicate that PACAP is a physiologically occurring peptide in pancreas and that PACAP, in a glucose-dependent manner, activates beta-cells presumably via a high affinity PACAP-selective receptor, raises [Ca2+]i by increasing the activity of L-type Ca2+ channels, and consequently stimulates insulin release. PACAP appears to be by far the most potent insulinotropic peptide known.
Sigma-Aldrich
Pituitary adenylate cyclase activating polypeptide-38