  • Search Results
  • Bacteriophages with potential for inactivation of fish pathogenic bacteria: survival, host specificity and effect on bacterial community structure.
Bacteriophages with potential for inactivation of fish pathogenic bacteria: survival, host specificity and effect on bacterial community structure.
Marine drugs (2011-12-14)
Carla Pereira, Yolanda J Silva, Ana L Santos, Angela Cunha, Newton C M Gomes, Adelaide Almeida
ABSTRACT
Phage therapy may represent a viable alternative to antibiotics to inactivate fish pathogenic bacteria. Its use, however, requires the awareness of novel kinetics phenomena not applied to conventional drug treatments. The main objective of this work was to isolate bacteriophages with potential to inactivate fish pathogenic bacteria, without major effects on the structure of natural bacterial communities of aquaculture waters. The survival was determined in marine water, through quantification by the soft agar overlay technique. The host specificity was evaluated by cross infection. The ecological impact of phage addition on the structure of the bacterial community was evaluated by DGGE of PCR amplified 16S rRNA gene fragments. The survival period varied between 12 and 91 days, with a higher viability for Aeromonas salmonicida phages. The phages of Vibrio parahaemolyticus and of A. salmonicida infected bacteria of different families with a high efficacy of plating. The specific phages of pathogenic bacteria had no detectable impact on the structure of the bacterial community. In conclusion, V. parahaemolyticus and A. salmonicida phages show good survival time in marine water, have only a moderated impact on the overall bacterial community structure and the desired specificity for host pathogenic bacteria, being potential candidates for therapy of fish infectious diseases in marine aquaculture systems.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
Bovine Serum Albumin, pH <5.0, powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, low endotoxin, lyophilized powder, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, For use as a marker in SDS-PAGE