  • Search Results
  • The p53-cathepsin axis cooperates with ROS to activate programmed necrotic death upon DNA damage.
The p53-cathepsin axis cooperates with ROS to activate programmed necrotic death upon DNA damage.
Proceedings of the National Academy of Sciences of the United States of America (2009-01-16)
Ho-Chou Tu, Decheng Ren, Gary X Wang, David Y Chen, Todd D Westergard, Hyungjin Kim, Satoru Sasagawa, James J-D Hsieh, Emily H-Y Cheng
ABSTRACT
Three forms of cell death have been described: apoptosis, autophagic cell death, and necrosis. Although genetic and biochemical studies have formulated a detailed blueprint concerning the apoptotic network, necrosis is generally perceived as a passive cellular demise resulted from unmanageable physical damages. Here, we conclude an active de novo genetic program underlying DNA damage-induced necrosis, thus assigning necrotic cell death as a form of "programmed cell death." Cells deficient of the essential mitochondrial apoptotic effectors, BAX and BAK, ultimately succumbed to DNA damage, exhibiting signature necrotic characteristics. Importantly, this genotoxic stress-triggered necrosis was abrogated when either transcription or translation was inhibited. We pinpointed the p53-cathepsin axis as the quintessential framework underlying necrotic cell death. p53 induces cathepsin Q that cooperates with reactive oxygen species (ROS) to execute necrosis. Moreover, we presented the in vivo evidence of p53-activated necrosis in tumor allografts. Current study lays the foundation for future experimental and therapeutic discoveries aimed at "programmed necrotic death."
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%