  • Search Results
  • Enhanced expression of the M-type phospholipase A2 receptor in glomeruli correlates with serum receptor antibodies in primary membranous nephropathy.
Enhanced expression of the M-type phospholipase A2 receptor in glomeruli correlates with serum receptor antibodies in primary membranous nephropathy.
Kidney international (2012-06-08)
Elion Hoxha, Ursula Kneißler, Gesa Stege, Gunther Zahner, Ina Thiele, Ulf Panzer, Sigrid Harendza, Udo M Helmchen, Rolf A K Stahl
ABSTRACT
The M-type phospholipase A2 receptor (PLA2R) is the major target antigen in idiopathic membranous nephropathy with detectable autoantibodies in the serum of up to 70% of patients. In retrospective studies, the PLA2R-autoantibody titer in the serum was sometimes negative indicating their measurement alone may be inconclusive. In order to better differentiate between primary and secondary membranous nephropathy, we conducted a prospective study that included 88 patients with a histologic diagnosis of membranous nephropathy. Immunohistochemical analysis for PLA2R was faintly positive in kidneys from normal individuals and patients with various other glomerular injuries. In 61 of the 88 patients, PLA2R expression was strongly positive in glomeruli, and in 60 of these patients PLA2R autoantibodies were also detected in the serum. The 27 patients negative for serum PLA2R autoantibodies were faintly positive for PLA2R staining in glomeruli and in 15 of these patients a secondary cause was found. The remaining 12 patients have a yet undetected secondary cause of membranous nephropathy or have different glomerular antigens other than PLA2R. Thus, increased staining for PLA2R in glomeruli of renal biopsies tightly correlates with the presence of PLA2R autoantibodies in the serum and this may help discriminate between primary and secondary membranous nephropathy.
Sigma-Aldrich
Anti-PLA2R1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Human IgG4−FITC antibody produced in mouse, clone HP-6025, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Human IgG3−FITC antibody produced in mouse, clone HP-6050, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG1−FITC antibody produced in mouse, clone 8c/6-39, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Human IgG2−FITC antibody produced in mouse, clone HP-6014, purified immunoglobulin, buffered aqueous solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.