  • Search Results
  • A human neoplastic hematopoietic cell line producing a fibroblast type of interferon.
A human neoplastic hematopoietic cell line producing a fibroblast type of interferon.
Developments in biological standardization (1979-01-01)
I Larsson, E Lundgren, K Nilsson, O Strannegard
PMID223921
ABSTRACT
1301 is a subline of the EBV-genome negative T leukemia line CCRF-CEM. IF was induced by Sendai virus. By use of neutralizing antisera and by studies of some physical properties it could be shown that IF was of the fibroblast type. This was confirmed by its antiviral effect on heterologous cell lines.
Product Description
1301 Cell Line human, 01051619, T-cell leukemia