  • Search Results
  • Productive, persistent infection of human colorectal cell lines with human immunodeficiency virus.
Productive, persistent infection of human colorectal cell lines with human immunodeficiency virus.
Journal of virology (1987-01-01)
A Adachi, S Koenig, H E Gendelman, D Daugherty, S Gattoni-Celli, A S Fauci, M A Martin
ABSTRACT
Thirteen adherent human non-lymphocyte cell lines were tested for their susceptibility to infection by human immunodeficiency virus. Productive infection could be demonstrated in three of five colorectal carcinoma cell lines examined; the other eight human non-lymphocyte cell lines were uninfectible. A susceptible colon carcinoma cell line (HT29), as well as normal colonic mucosa, was shown to contain a 3.0-kilobase species of poly(A)+ CD4 RNA, whereas uninfectible colon carcinoma and rhabdomyosarcoma cell lines synthesized no detectable T4 RNA. A persistently infected colon carcinoma cell line was established that continued to produce progeny human immunodeficiency virus for more than 10 weeks postinfection.
Sigma-Aldrich
CACO-2, Caucasian colon adenocarcinoma
CACO-2 Cell Line human, 86010202, from human colon(Caucasian colon adenocarcinoma)