  • Search Results
  • Absence of HeLa cell contamination in 169 cell lines derived from human tumors.
Absence of HeLa cell contamination in 169 cell lines derived from human tumors.
Journal of the National Cancer Institute (1977-02-01)
J Fogh, W C Wright, J D Loveless
PMID833871
ABSTRACT
Numerous cell lines derived from human tumors are not HeLa contaminants. Of 192 lines established in this or other laboratories, 169 lines were found to be G6PD type B. Twenty-three lines were type A as HeLa; three of these were of Negroid origin. There is reasonable doubt that the remaining 20 lines will all be shown to be confounded with HeLa.
Sigma-Aldrich
CACO-2, Caucasian colon adenocarcinoma
CACO-2 Cell Line human, 86010202, from human colon(Caucasian colon adenocarcinoma)
SK-OV-3 Cell Line human, 91091004
SAOS-2 Cell Line human, primary osteogenic sarcoma, 89050205
MDA-MB-361, 92020423
CAKI 2, 93120819
CALU 1, 93120818
SK MES 1, 93120837
SK-HEP-1, 91091816
SW 626, 91091203