  • Search Results
  • Staining diatoms with rhodamine dyes: control of emission colour in photonic biocomposites.
Staining diatoms with rhodamine dyes: control of emission colour in photonic biocomposites.
Journal of the Royal Society, Interface (2011-08-26)
Melanie Kucki, Thomas Fuhrmann-Lieker
ABSTRACT
The incorporation of rhodamine dyes in the cell wall of diatoms Coscinodiscus granii and Coscinodiscus wailesii for the production of luminescent hybrid nanostructures is investigated. By systematic variation of the substitution pattern of the rhodamine core, we found that carbonic acids are considerably better suited than esters because of their physiological compatibility. The amino substitution pattern that controls the optical properties of the chromophore has no critical influence on dye uptake and incorporation, thus a variety of biocomposites with different emission maxima can be prepared. Applications in biomineralization studies as well as in materials science are envisioned.
Sigma-Aldrich
Atto Rho13 NHS ester
Sigma-Aldrich
Atto Rho13 maleimide
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.