  • Search Results
  • Perinatal undernutrition affects the methylation and expression of the leptin gene in adults: implication for the understanding of metabolic syndrome.
Perinatal undernutrition affects the methylation and expression of the leptin gene in adults: implication for the understanding of metabolic syndrome.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2011-06-15)
Céline Jousse, Laurent Parry, Sarah Lambert-Langlais, Anne-Catherine Maurin, Julien Averous, Alain Bruhat, Valérie Carraro, Jorg Tost, Philippe Letteron, Patty Chen, Ralf Jockers, Jean-Marie Launay, Jacques Mallet, Pierre Fafournoux
ABSTRACT
Transient environmental influences, such as perinatal nutritional stress, may induce deleterious metabolic symptoms that last for the entire life of individuals, implying that epigenetic modifications play an important role in this process. We have investigated, in mice, the consequences of maternal undernutrition during gestation and lactation on DNA methylation and expression of the leptin gene, which plays a major regulatory role in coordinating nutritional state with many aspects of mammalian biology. We show that animals born to mothers fed a low-protein-diet (F1-LPD group) have a lower body weight/adiposity and exhibit a higher food intake than animals born to mothers fed a control diet (F1-CD group). These modifications persisted throughout life and were associated with lower levels of leptin mRNA and protein in starved F1-LPD mice, emphasizing that maternal protein-undernutrition affects the balance between food intake and energy expenditure in adults. Moreover, this nutritional stress resulted in the removal of methyls at CpGs located in the promoter of leptin, causing a permanent specific modification in the dynamics of the expression of leptin, which exhibits a stronger induction in the F1-LPD than in F1-CD mice in response to a meal. This study is an example of a molecular rationale linking transient environmental influences to permanent phenotypic consequences.
Sigma-Aldrich
Imprint® Methylated DNA Quantification Kit, To measure global DNA methylation shifts from as low as 10 ng DNA