  • Search Results
  • Salicylic acid is important for basal defense of Solanum tuberosum against Phytophthora infestans.
Salicylic acid is important for basal defense of Solanum tuberosum against Phytophthora infestans.
Molecular plant-microbe interactions : MPMI (2007-11-06)
Vincentius A Halim, Lennart Eschen-Lippold, Simone Altmann, Mandy Birschwilks, Dierk Scheel, Sabine Rosahl
ABSTRACT
The importance of the signaling compound salicylic acid for basal defense of potato (Solanum tuberosum L. cv. Désirée) against Phytophthora infestans, the causal agent of late blight disease, was assessed using transgenic NahG potato plants which are unable to accumulate salicylic acid. Although the size of lesions caused by P. infestans was not significantly different in wild-type and transgenic NahG plants, real-time polymerase chain reaction analyses revealed a drastic enhancement of pathogen growth in potato plants depleted of salicylic acid. Increased susceptibility of NahG plants correlated with compromised callose formation and reduced early defense gene expression. NahG plants pretreated with the salicylic acid analog 2,6-dichloro-isonicotinic acid allowed pathogen growth to a similar extent as did wild-type plants, indicating that salicylic acid is an important compound required for basal defense of potato against P. infestans.
Sigma-Aldrich
2,6-Dichloropyridine-4-carboxylic acid, 98%